Skip to Content
Merck
All Photos(1)

Key Documents

548804

Sigma-Aldrich

Ytterbium

chunks, 99.9% trace metals basis

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Yb
CAS Number:
Molecular Weight:
173.04
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.23

Assay

99.9% trace metals basis

form

chunks

reaction suitability

reagent type: catalyst
core: ytterbium

resistivity

28 μΩ-cm, 20°C

bp

1196 °C (lit.)

mp

824 °C (lit.)

density

6.54 g/mL at 25 °C (lit.)

SMILES string

[Yb]

InChI

1S/Yb

InChI key

NAWDYIZEMPQZHO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zhen Liu et al.
Biomaterials, 34(30), 7444-7452 (2013-07-16)
Nanoparticulate contrast agents have attracted a great deal of attention along with the rapid development of modern medicine. Here, a binary contrast agent based on PAA modified BaYbF5:Tm nanoparticles for direct visualization of gastrointestinal (GI) tract has been designed and
Jianhua Wang et al.
Optics express, 20(27), 28373-28378 (2012-12-25)
A 100 W-class all-fiber linearly-polarized single-mode fiber laser at 1120 nm with an optical efficiency of 50% was demonstrated. The laser consists of a 4.2 m long Yb-doped polarization maintaining fiber with a core diameter of 10 μm and a
S Rydberg et al.
Optics express, 21(6), 6681-6688 (2013-04-03)
In this work we present experimental evidence that the valence instability of the ytterbium ion play a key role for the observed photodarkening mechanism in Yb-doped fiber lasers. Luminescence and excitation spectroscopy performed on UV irradiated Yb/Al doped silica glass
Luc Beaulieu et al.
Medical physics, 39(10), 6208-6236 (2012-10-09)
The charge of Task Group 186 (TG-186) is to provide guidance for early adopters of model-based dose calculation algorithms (MBDCAs) for brachytherapy (BT) dose calculations to ensure practice uniformity. Contrary to external beam radiotherapy, heterogeneity correction algorithms have only recently
A E Bednyakova et al.
Optics express, 21(7), 8177-8182 (2013-04-11)
We present direct real-time experimental measurements and numerical modeling of temporal and statistical properties for the Ytterbium-doped fiber laser with spectral bandwidth of ~2 GHz. The obtained results demonstrate nearly exponential probability density function for intensity fluctuations. A significant decrease

Articles

A significant limiting factor for wearable electronics and wireless sensors is the finite amount of energy that can be stored in on-board batteries.

Rechargeable solid-state batteries are becoming increasingly important due to wide-spread use in computers, portable electronics, and vehicular applications.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service