544329
7-Ethoxy-4-methylcoumarin
98%
Synonym(s):
Ethyl 4-methylumbelliferyl ether
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C12H12O3
CAS Number:
Molecular Weight:
204.22
Beilstein:
169998
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
Recommended Products
Assay
98%
mp
114-116 °C (lit.)
SMILES string
CCOc1ccc2C(C)=CC(=O)Oc2c1
InChI
1S/C12H12O3/c1-3-14-9-4-5-10-8(2)6-12(13)15-11(10)7-9/h4-7H,3H2,1-2H3
InChI key
NKRISXMDKXBVRJ-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
General description
7-Ethoxy-4-methylcoumarin (EtOMC), also known as ethyl 4-methylumbelliferyl ether, is a coumarin derivative. Its standard molar energy of combustion is ?5888.0 ± 3.2kJ·mol?1 and standard molar enthalpy of formation in the crystalline phase is ?545.4 ± 3.6kJ·mol?1.EtOMC can be prepared by reacting methyl acetoacetate with 3-ethoxyphenol in the presence of boron trifluoride dihydrate.The fluorescence quenching of EtOMC is more effective in the presence 4-hydroxy-TEMPO (4-hydroxy-2,2,6,6-tetramethylpiperidin-1-oxyl).
Application
7-Ethoxy-4-methylcoumarin may be used in the assay of 4-chlormethyl-7-ethoxycoumarin O-deethylase activity.
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Standard molar enthalpies of formation in the crystalline phase of 7-hydroxy-4-methylcoumarin, 7-ethoxy-4-methylcoumarin, and 6-methoxy-4-methylcoumarin.
Amador P, et al.
The Journal of Chemical Thermodynamics, 43(9), 1414-1416 (2011)
Analysis of fluorescence quenching of coumarin derivatives by 4-hydroxy-TEMPO in aqueous solution.
Zamojc K, et al.
Journal of Fluorescence, 24(3), 713-718 (2014)
U D Kuhn et al.
Experimental and toxicologic pathology : official journal of the Gesellschaft fur Toxikologische Pathologie, 50(4-6), 491-496 (1998-10-23)
7-Ethoxycoumarin (EC) is widely used as a model substrate for monooxygenase function, its O-deethylation representing cytochrome P450 (P450) activity mainly of 1A but also of 2B isoforms. Reports on investigations of its own capacity to induce or suppress P450 activities
Rongjing Yang et al.
Se pu = Chinese journal of chromatography, 30(2), 160-164 (2012-06-12)
A rapid analytical method for the determination of five coumarins (coumarin, 7-methoxycoumarin, dihydrocoumarin, 7-methyl coumarin and 7-ethoxy-4-methyl coumarin) in toys by high performance liquid chromatography-tandem mass spectrometry (HPLC-MS/MS) has been developed. After ultrasonic extraction in tetrahydrofuran, the samples were analyzed
S Ekins et al.
The Journal of pharmacology and experimental therapeutics, 286(3), 1253-1259 (1998-09-11)
Previous studies in this laboratory have determined the lack of specificity of several antibody and substrate probes of CYP2B6. The goals of the current study were to examine the expression of CYP2B6 in a bank of human liver microsome (HLM)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service