Skip to Content
Merck
All Photos(1)

Key Documents

531456

Sigma-Aldrich

5-Chloro-2-furaldehyde

98%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H3ClO2
CAS Number:
Molecular Weight:
130.53
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

mp

34-37 °C (lit.)

functional group

aldehyde
chloro

storage temp.

2-8°C

SMILES string

Clc1ccc(C=O)o1

InChI

1S/C5H3ClO2/c6-5-2-1-4(3-7)8-5/h1-3H

InChI key

DGAUAVDWXYXXGQ-UHFFFAOYSA-N

General description

5-Chloro-2-furaldehyde is also known as 5-chlorofurfural. It reacts with aniline and aniline hydrochloride to form bis-(phenylamino) derivatives, without the furan ring cleavage.[1] It also undergoes coupling with ethyl acetoacetate to form the corresponding ethyl bis-acetoacetate.[2]

Application

5-Chloro-2-furaldehyde may be used as an internal standard during the analysis of furanic compounds by reversed-phase-high performance liquid chromatography–diode array detection (RP-HPLC–DAD) method.[3]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The Synthesis of Substituted ?-Thienyl-and ?-Furylglutaric Acids.
Smith Jr WT and Shelton RW.
Journal of the American Chemical Society, 76(10), 2731-2732 (1954)
Waterpipe smoke: a considerable source of human exposure against furanic compounds.
Schubert J, et al.
Analytica Chimica Acta, 709, 105-112 (2012)
Utilization of potassium vinyltrifluoroborate in the development of a 1, 2-dianion equivalent.
Molander GA and Sandrock Deidre L
Organic Letters, 11(11), 2369-2372 (2009)
?-Selective directed catalytic asymmetric hydroboration of 1,1-disubstituted alkenes.
Smith SM, et al.
Chemical Communications (Cambridge, England), 48(100), 12180-12182 (2012)
The Reaction of Certain Substituted Furfurals with Aniline and Aniline Hydrochloride.
Drisko RW and McKennis Jr H.
Journal of the American Chemical Society, 74(10), 2626-2628 (1952)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service