Skip to Content
Merck
All Photos(1)

Key Documents

528498

Sigma-Aldrich

3-Ethoxybenzoic acid

97%

Sign Into View Organizational & Contract Pricing

Select a Size

10 G
CZK 2,210.00

CZK 2,210.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
10 G
CZK 2,210.00

About This Item

Linear Formula:
C2H5OC6H4CO2H
CAS Number:
Molecular Weight:
166.17
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

CZK 2,210.00


Please contact Customer Service for Availability

Request a Bulk Order

Assay

97%

mp

136-140 °C (lit.)

functional group

carboxylic acid

SMILES string

CCOc1cccc(c1)C(O)=O

InChI

1S/C9H10O3/c1-2-12-8-5-3-4-7(6-8)9(10)11/h3-6H,2H2,1H3,(H,10,11)

InChI key

DTFQMPQJMDEWKJ-UHFFFAOYSA-N

General description

3-Ethoxybenzoic acid is a xenobiotic compound.[1]

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Katsushi Amano et al.
Journal of immunology (Baltimore, Md. : 1950), 174(9), 5874-5883 (2005-04-23)
Emerging evidence has suggested environmental factors as causative agents in the pathogenesis of primary biliary cirrhosis (PBC). We have hypothesized that in PBC the lipoyl domain of the immunodominant E2 component of pyruvate dehydrogenase (PDC-E2) is replaced by a chemical

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service