Skip to Content
Merck
All Photos(1)

Key Documents

522457

Sigma-Aldrich

3-Amino-5-nitrobenzisothiazole

97%

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
CZK 990.00

CZK 990.00

List PriceCZK 1,980.00Save 50%

Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
5 G
CZK 990.00

About This Item

Empirical Formula (Hill Notation):
C7H5N3O2S
CAS Number:
Molecular Weight:
195.20
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

CZK 990.00

List PriceCZK 1,980.00Save 50%

Please contact Customer Service for Availability

Request a Bulk Order

Quality Level

Assay

97%

mp

250 °C (dec.) (lit.)

functional group

nitro

SMILES string

Nc1nsc2ccc(cc12)[N+]([O-])=O

InChI

1S/C7H5N3O2S/c8-7-5-3-4(10(11)12)1-2-6(5)13-9-7/h1-3H,(H2,8,9)

InChI key

LDTCWISGJYTXDC-UHFFFAOYSA-N

General description

3-Amino-5-nitrobenzisothiazole forms 1:1 complex with β-cyclodextrin (β-CDx). Structural and photoprototropic features of this complex have been investigated.[1][2]

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

R Rajamohan et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 69(2), 371-377 (2007-07-03)
The inclusion complexation and photoprototropic behaviour of 3-amino-5-nitrobenzisothiazole (ANBT) in aqueous beta-cyclodextrin (beta-CDx) solution have been investigated. Absorption and fluorescence intensities of the neutral form of ANBT are enhanced due to the formation of 1:1 complex with beta-CDx. The complex
Theoretical study of inclusion complexation of 3-amino-5-nitrobenzisothiazole with ?-cyclodextrin.
Leila N, et al.
Journal of Molecular Liquids, 160(1), 8-13 (2011)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service