510343
(+)-N-(1(S)-Phenylethyl)-1-azabicyclo[2.2.2]octan-3(R)-amine hydrochloride
98%
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C15H22N2 · 2HCl
CAS Number:
Molecular Weight:
303.27
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
Assay
98%
optical activity
[α]20/D +2°, c = 2 in H2O
mp
285 °C (dec.) (lit.)
SMILES string
Cl.Cl.C[C@H](N[C@H]1CN2CC[C@H]1CC2)c3ccccc3
InChI
1S/C15H22N2.2ClH/c1-12(13-5-3-2-4-6-13)16-15-11-17-9-7-14(15)8-10-17;;/h2-6,12,14-16H,7-11H2,1H3;2*1H/t12-,15-;;/m0../s1
InChI key
BABMCKVVGDIUCA-XPTKPXFUSA-N
General description
May contain up to 2.5% monohydrochloride salt
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service