Skip to Content
Merck
All Photos(3)

Key Documents

495786

Sigma-Aldrich

Perfluorinated resin, powder made from Nafion

greener alternative

Synonym(s):

Perfluoro-3,6-dioxa-4-methyl-7-octene-1-sulfonic acid, Tetrafluoroethylene-perfluoro-3,6-dioxa-4-methyl-7-octenesulfonic acid copolymer

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
26111700
eCl@ss:
32110203
NACRES:
NA.23

form

powder

eq. wt.

1,100

greener alternative product characteristics

Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

particle size

35-60 mesh

mp

>350 °C (lit.)

greener alternative category

InChI

1S/C7HF13O5S.C2F4/c8-1(9)2(10)24-5(15,16)3(11,4(12,13)14)25-6(17,18)7(19,20)26(21,22)23;3-1(4)2(5)6/h(H,21,22,23);

InChI key

FOYUGSIADQEOEK-UHFFFAOYSA-N

General description

We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for energy efficiency. Find details here.

Application

Perfluorinated resin, powder made from Nafion can be used in the formation of proton exchange membranes. Its properties include high proton conductivity and good physio-chemical properties. It can further be used in the fabrication of polymeric fuel cells.

Physical form

Hydrogen ion form.

Legal Information

Nafion is a trademark of The Chemours Company FC, LLC

Storage Class Code

11 - Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Polyazomethines and their acid-base interactions with Nafion and Nafion-imidazole membranes for efficient fuel cells
Malinowski M, et al.
Sustainable Energy & Fuels, 1(8), 1810-1819 (2017)
Studies of bibenzimidazole and imidazole influence on electrochemical properties of polymer fuel cells
Iwan A, et al.
Electrochimica Acta, 164, 143-153 (2015)
Min-Ho Park et al.
Advanced materials (Deerfield Beach, Fla.), 27(29), 4308-4314 (2015-06-24)
A novel flexible encapsulation method (Flex Lami-capsulation) is reported, which can be applied in the roll-to-roll process for mass production of organic electronic devices. Flex Lami-capsulation is very simple, fast, and getter-free, and is as effective as glass encapsulation. Use

Articles

Proton exchange membrane (PEM) fuel cells operate at relatively low temperatures and are composed of two electrodes and a conductive elecrolyte.

Advances in electrochemical water conversion and understanding PEMFC degradation drive progress in hydrogen technologies.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service