Skip to Content
Merck
All Photos(1)

Key Documents

493120

Sigma-Aldrich

Sarcosine-d3 (methyl-d3)

99 atom % D

Synonym(s):

N-Methyl-d3-glycine

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
CZK 12,620.00

CZK 12,620.00

List PriceCZK 63,100.00Save 80%

Please contact Customer Service for Availability

Request more information

Select a Size

Change View
1 G
CZK 12,620.00

About This Item

Linear Formula:
CD3NHCH2CO2H
CAS Number:
Molecular Weight:
92.11
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.12

CZK 12,620.00

List PriceCZK 63,100.00Save 80%

Please contact Customer Service for Availability

Request more information

isotopic purity

99 atom % D

Quality Level

form

solid

mp

208-212 °C

mass shift

M+3

SMILES string

[2H]C([2H])([2H])NCC(O)=O

InChI

1S/C3H7NO2/c1-4-2-3(5)6/h4H,2H2,1H3,(H,5,6)/i1D3

InChI key

FSYKKLYZXJSNPZ-FIBGUPNXSA-N

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service