Skip to Content
Merck
All Photos(1)

Key Documents

492582

Sigma-Aldrich

Ethylamine-15N hydrochloride

99 atom % 15N

Sign Into View Organizational & Contract Pricing

Select a Size

500 MG
CZK 17,100.00

CZK 17,100.00

List priceCZK 18,000.00Save 5%

Please contact Customer Service for Availability

Request more information

Select a Size

Change View
500 MG
CZK 17,100.00

About This Item

Linear Formula:
C2H515NH2·HCl
CAS Number:
Molecular Weight:
82.54
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.12

CZK 17,100.00

List priceCZK 18,000.00Save 5%

Please contact Customer Service for Availability

Request more information

isotopic purity

99 atom % 15N

form

solid

mp

107-108 °C (lit.)

mass shift

M+1

SMILES string

Cl.CC[15NH2]

InChI

1S/C2H7N.ClH/c1-2-3;/h2-3H2,1H3;1H/i3+1;

InChI key

XWBDWHCCBGMXKG-FJUFCODESA-N

Related Categories

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service