Skip to Content
Merck
All Photos(1)

Key Documents

490598

Sigma-Aldrich

Propane-1,1,1,3,3,3-d6

98 atom % D

Synonym(s):

1,1,1,3,3,3-Hexadeuteriopropane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CD3CH2CD3
CAS Number:
Molecular Weight:
50.13
MDL number:
UNSPSC Code:
12142201
PubChem Substance ID:
NACRES:
NA.12
Product 490598 is not currently sold in your country. Contact Technical Service

vapor density

1.5 (vs air)

Quality Level

vapor pressure

8.42 atm ( 21.1 °C)

isotopic purity

98 atom % D

bp

−42.1 °C (lit.)

mp

−188 °C (lit.)

mass shift

M+6

SMILES string

[2H]C([2H])([2H])CC([2H])([2H])[2H]

InChI

1S/C3H8/c1-3-2/h3H2,1-2H3/i1D3,2D3

InChI key

ATUOYWHBWRKTHZ-WFGJKAKNSA-N

Related Categories

Packaging

Standard 500 mL quantity is packaged in a 25 mL Sure/Pac cylinder with brass 1/4" male NPT valve. Nominal gas pressure at 21ºC is 100 psig for 500 mL. This pressure is slightly above atmospheric pressure. Care must be taken when extracting this product from the cylinder.
This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Recommended products

Brass hose adapter Z146811 or brass body mini gas regulator Z513539 is recommended.

Legal Information

Sure/Pac is a trademark of Sigma-Aldrich Co. LLC

hose barb

Product No.
Description
Pricing

regulator

Product No.
Description
Pricing

Pictograms

FlameGas cylinder

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Flam. Gas 1 - Press. Gas Compr. Gas

Storage Class Code

2A - Gases

WGK

nwg

Flash Point(F)

-155.2 °F - closed cup

Flash Point(C)

-104.00 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service