Skip to Content
Merck
All Photos(2)

Documents

483729

Sigma-Aldrich

mono-2-(Methacryloyloxy)ethyl maleate

Synonym(s):

(Z)-4-(2-(Methacryloyloxy)ethoxy)-4-oxobut-2-enoic acid, 2-Butenedioic Acid (Z)-Monophenyl Ester, Ethylene glycol methacrylate maleate, Mono-2-(methacryloyloxy)ethyl maleate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2C=C(CH3)CO2CH2CH2O2CCH=CHCO2H
CAS Number:
Molecular Weight:
228.20
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

contains

180-480 ppm monomethyl ether hydroquinone as inhibitor

refractive index

n20/D 1.483 (lit.)

density

1.217 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

CC(=C)C(=O)OCCOC(=O)\C=C/C(O)=O

InChI

1S/C10H12O6/c1-7(2)10(14)16-6-5-15-9(13)4-3-8(11)12/h3-4H,1,5-6H2,2H3,(H,11,12)/b4-3-

InChI key

HSFXEOPJXMFQHG-ARJAWSKDSA-N

Related Categories

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B - Skin Sens. 1

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

T Fukushima et al.
Journal of dental research, 69(1), 46-50 (1990-01-01)
Three types of methacryloyloxyalkyl hydrogen maleates--2-(methacryloyloxy)ethyl hydrogen maleate (2MEM), 1-(methacryloyloxy-methyl)ethyl hydrogen maleate (MMEM), and 1-(methacryloyloxymethyl)propyl hydrogen maleate (MMPM)--were prepared for use as bonding agents. The relationships between the thickness of an acid-proof dentin layer and the bond strength and chemical
S Venz et al.
Journal of dental research, 72(3), 582-586 (1993-03-01)
PMGDM, a PMDM-type adhesive monomer, was synthesized from pyromellitic dianhydride and glycerol dimethacrylate. Only the para isomer of PMDM, a solid, is used in dental adhesives. The adhesive monomer PMGDM is a liquid and consists of a mixture of para-

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service