Skip to Content
Merck
All Photos(2)

Key Documents

480088

Sigma-Aldrich

3,5-Dimethylphenylboronic acid

≥95.0%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2C6H3B(OH)2
CAS Number:
Molecular Weight:
149.98
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

Assay

≥95.0%

mp

261-265 °C (lit.)

SMILES string

Cc1cc(C)cc(c1)B(O)O

InChI

1S/C8H11BO2/c1-6-3-7(2)5-8(4-6)9(10)11/h3-5,10-11H,1-2H3

InChI key

DJGHSJBYKIQHIK-UHFFFAOYSA-N

Application

3,5-Dimethylphenylboronic acid (DMPBA) can be used as:
  • A reactant in the palladium-catalyzed Suzuki coupling reactions.
  • A co-extractant in the combination with modified Aliquat 336 for the extraction of xylose, glucose, and fructose from aqueous solutions.
  • A reactant to prepare penultimate methyl ester.

Other Notes

Contains varying amounts of anhydride

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

SAR studies of novel 5-substituted 2-arylindoles as nonpeptidyl GnRH receptor antagonists
Chu L, et al.
Bioorganic & medicinal chemistry letters, 11(4), 515-517 (2001)
Purification and Concentration of Xylose and Glucose from Neutralized Bagasse Hydrolysates Using 3, 5-Dimethylphenylboronic Acid and Modified Aliquat 336 as Coextractants
Griffin, GJ
Separation Science and Technology, 40(11), 2337-2351 (2005)
Suzuki reaction of a diarylborinic acid: one-pot preparation and cross-coupling of bis (3, 5-dimethylphenyl) borinic acid
Winkle DD and Schaab KM
Organic Process Research & Development, 5(4), 450-451 (2001)
Guillaume Erbland et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 25(71), 16328-16339 (2019-10-12)
The design and synthesis of two families of molecular-gear prototypes is reported, with the aim of assembling them into trains of gears on a surface and ultimately achieving controlled intermolecular gearing motion. These piano-stool ruthenium complexes incorporate a hydrotris(indazolyl)borate moiety

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service