Skip to Content
Merck
All Photos(1)

Key Documents

47306

Sigma-Aldrich

m-Fluoro-L-phenylalanine

≥99.0% (sum of enantiomers, HPLC)

Synonym(s):

3-Fluoro-L-phenylalanine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H10FNO2
CAS Number:
Molecular Weight:
183.18
Beilstein:
2725762
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.22

Assay

≥99.0% (sum of enantiomers, HPLC)

optical activity

[α]20/D −25±2°, c = 1% in H2O

reaction suitability

reaction type: solution phase peptide synthesis

application(s)

peptide synthesis

SMILES string

N[C@@H](Cc1cccc(F)c1)C(O)=O

InChI

1S/C9H10FNO2/c10-7-3-1-2-6(4-7)5-8(11)9(12)13/h1-4,8H,5,11H2,(H,12,13)/t8-/m0/s1

InChI key

VWHRYODZTDMVSS-QMMMGPOBSA-N

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

N Rastogi et al.
Antimicrobial agents and chemotherapy, 34(11), 2061-2064 (1990-11-01)
In the present work, we investigated whether resistance to isoniazid (INH) of organisms belonging to the Mycobacterium avium complex was caused by the bacterial cell envelope, with the cell wall and the outer layer acting as an exclusion barrier. We
T T Otani et al.
Research communications in chemical pathology and pharmacology, 40(2), 321-324 (1983-05-01)
The fluoro-, chloro-, methoxy- and nitro-substituted benzoyl derivatives of m-fluoro-DL-phenylalanine, substituted singly at the ortho, meta or para position of the benzoyl phenyl ring, were prepared and tested for growth-inhibitory activity in a Lactobacillus casei system used as an antitumor
K Fukuda et al.
Current genetics, 20(6), 453-456 (1991-12-01)
o-Fluoro-DL-phenylalanine (OFP)-resistant mutants which overproduce beta-phenethyl-alcohol were isolated from a laboratory strain of Saccharomyces cerevisiae. Cells of one of the mutants accumulated tyrosine and phenylalanine 1.5-3 fold more than did wild-type cells. Its 3-deoxy-D-arabino-hepturosonate-7-phosphate (DAHP) synthase (EC 4.1.2.15), encoded by
W D Kingsbury et al.
Journal of medicinal chemistry, 26(12), 1725-1729 (1983-12-01)
A series of di- and tripeptides containing D- and L-m-fluorophenylalanine was prepared and tested in vitro for the ability to inhibit the growth of the yeast Candida albicans. The results demonstrate that peptides containing L-m-fluorophenylalanine inhibited the growth of C.
[Transport of phenylalanine in human brain by L-[2-18F]-fluorophenylalanine and PET].
S Miura et al.
Nihon rinsho. Japanese journal of clinical medicine, 49(7), 1537-1540 (1991-07-01)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service