Skip to Content
Merck
All Photos(2)

Key Documents

47297

Sigma-Aldrich

6-(Fmoc-amino)-1-hexanol

≥98.0%

Synonym(s):

9-Fluorenylmethyl N-(6-hydroxyhexyl)carbamate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H25NO3
CAS Number:
Molecular Weight:
339.43
Beilstein:
4531101
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.22

Assay

≥98.0%

reaction suitability

reagent type: cross-linking reagent

functional group

Fmoc
hydroxyl

storage temp.

2-8°C

SMILES string

OCCCCCCNC(OCC1C(C=CC=C2)=C2C3=C1C=CC=C3)=O

InChI

1S/C21H25NO3/c23-14-8-2-1-7-13-22-21(24)25-15-20-18-11-5-3-9-16(18)17-10-4-6-12-19(17)20/h3-6,9-12,20,23H,1-2,7-8,13-15H2,(H,22,24)

InChI key

VGXOJZUTHIGHPT-UHFFFAOYSA-N

Other Notes

Reagent for the introduction of a spacer arm.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S Bay et al.
Carbohydrate research, 298(3), 153-161 (1997-03-05)
The T-antigen [beta-D-Gal-(1-->3)-D-Ga1NAc] has been linked to biotin through a C6 spacer arm for the detection of a specific 'T-antigen-lectin' complex at the surface and/or on the migration pathway of melanoma cells. When 4,6-di-O-acetyl-2-azido-2-deoxy-3-O-(2,3,4, 6-tetra-O-acetyl-beta-D-galactopyranosyl)-alpha- or -beta-D-galactopyranosyl halides were treated
S. Agrawal
Tetrahedron Letters, 30, 7025-7025 (1989)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service