Skip to Content
Merck
All Photos(2)

Key Documents

465089

Sigma-Aldrich

3,4-Difluorophenylboronic acid

≥95%

Synonym(s):

3,4-Difluorobenzeneboronic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
F2C6H3B(OH)2
CAS Number:
Molecular Weight:
157.91
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

Assay

≥95%

mp

305-310 °C (lit.)

SMILES string

OB(O)c1ccc(F)c(F)c1

InChI

1S/C6H5BF2O2/c8-5-2-1-4(7(10)11)3-6(5)9/h1-3,10-11H

InChI key

RMGYQBHKEWWTOY-UHFFFAOYSA-N

Application

3,4-Difluorophenylboronic acid can be used as a reactant to prepare:
  • Fluorinated biaryl derivatives via Suzuki cross-coupling reaction with aryl and heteroaryl halides.
  • Flurodiarylmethanols by reacting with aryl aldehydes using Ni catalyst.
  • Conjugated fluorodiazaborinines by treating with diamines via intermolecular dehydration reaction for the detection of explosives.

Reactant involved in:
  • Suzuki cross-coupling reactions with aryl and heteroaryl halides
  • Oxo directing Liebeskind-Srogl cross-coupling reactions with gem-dihaloolefin-type α-oxo ketene dithioacetals
  • Substitution reactions with enyne acetates and carbonates

Other Notes

Contains varying amounts of anhydride

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Green synthesis of fluorinated biaryl derivatives via thermoregulated ligand/palladium-catalyzed Suzuki reaction
L Ning, et al.
Journal of Organometallic Chemistry, 696(13), 2641-2647 (2011)
NBN-Doped Conjugated Polycyclic Aromatic Hydrocarbons as an AIEgen Class for Extremely Sensitive Detection of Explosives
Wan W-M, et al.
Angewandte Chemie (International Edition in English), 57(47), 15510-15516 (2018)
Nickel salt-catalyzed addition reaction of arylboronic acids to aromatic aldehydes
Zhou L, et al.
Tetrahedron Letters, 50(4), 406-408 (2009)
Mingyan Jia et al.
Food chemistry, 335, 127566-127566 (2020-08-04)
In this work, we developed an optical colorimetric sensor array for the discrimination of Chinese teas. The sensor array was carefully designed based on tea polyphenol induced indicators displacement assay (IDA), using phenylboronic acids with different substituents as the receptors

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service