Skip to Content
Merck
All Photos(1)

Key Documents

446858

Sigma-Aldrich

2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,11,11,11-Heneicosafluoro-1-undecanol

95%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CF3(CF2)9CH2OH
CAS Number:
Molecular Weight:
550.11
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

95%

form

solid

bp

143 °C/80 mmHg (lit.)

mp

102-105 °C (lit.)

functional group

fluoro
hydroxyl

SMILES string

OCC(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)F

InChI

1S/C11H3F21O/c12-2(13,1-33)3(14,15)4(16,17)5(18,19)6(20,21)7(22,23)8(24,25)9(26,27)10(28,29)11(30,31)32/h33H,1H2

InChI key

MPAMAEMOVKLPRB-UHFFFAOYSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Evidence of an odd-even effect on the thermodynamic parameters of odd fluorotelomer alcohols.
Costa JCS, et al.
The Journal of Chemical Thermodynamics, 54, 171-178 (2012)
J Jackson Ellington et al.
Journal of chromatography. A, 1216(28), 5347-5354 (2009-06-06)
This article describes the development of an analytical method for the determination of fluorotelomer alcohols (FTOHs) in soil. The sensitive and selective determination of the telomer alcohols was performed by extraction with methyl tert-butyl ether (MTBE) and analysis of the

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service