Skip to Content
Merck
All Photos(1)

Key Documents

446505

Sigma-Aldrich

2-Fluoro-6-hydroxybenzoic acid

97%

Synonym(s):

6-Fluorosalicylic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
FC6H3(OH)CO2H
CAS Number:
Molecular Weight:
156.11
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

mp

159-163 °C (lit.)

SMILES string

OC(=O)c1c(O)cccc1F

InChI

1S/C7H5FO3/c8-4-2-1-3-5(9)6(4)7(10)11/h1-3,9H,(H,10,11)

InChI key

BCEKGWWLVKXZKK-UHFFFAOYSA-N

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Juliane Korp et al.
Chembiochem : a European journal of chemical biology, 16(17), 2445-2450 (2015-09-30)
The siderophore myxochelin A is a potent inhibitor of human 5-lipoxygenase (5-LO). To clarify whether the iron-chelating properties of myxochelin A are responsible for this activity, several analogues of this compound were generated in the native producer Pyxidicoccus fallax by precursor-directed biosynthesis.
Inhibitors of hepatitis C virus polymerase: synthesis and characterization of novel 2-oxy-6-fluoro-N-((S)-1-hydroxy-3-phenylpropan-2-yl)-benzamides.
Cheng CC, et al.
Bioorganic & Medicinal Chemistry Letters, 20(7), 2119-2124 (2010)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service