Skip to Content
Merck
All Photos(2)

Key Documents

441635

Sigma-Aldrich

3-Ethoxyphenylboronic acid

Synonym(s):

3-Ethoxybenzeneboronic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C2H5OC6H4B(OH)2
CAS Number:
Molecular Weight:
165.98
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

form

fibers

impurities

<15% anhydride

mp

139-146 °C (lit.)

SMILES string

CCOc1cccc(c1)B(O)O

InChI

1S/C8H11BO3/c1-2-12-8-5-3-4-7(6-8)9(10)11/h3-6,10-11H,2H2,1H3

InChI key

CHCWUTJYLUBETR-UHFFFAOYSA-N

Application

Reactant involved in:
  • Cyanation for synthesis of aromatic and vinyl nitriles
  • Microwave-assisted palladium-catalyzed arylation of resin supported pyrazinones
  • Copper-mediated N- and O-arylations

Reactant involved in synthesis of biologically active molecules including:
  • Amino derivatives of indole for use as isoprenylcysteine carboxyl methyltransferase inhibitors
  • 5-Arylindazole glucocorticoid receptor agonists and antagonists
  • 1,5-Diaryl-1,2,4-triazoles as cis-restricted combretastatin analogs

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mingyan Jia et al.
Food chemistry, 335, 127566-127566 (2020-08-04)
In this work, we developed an optical colorimetric sensor array for the discrimination of Chinese teas. The sensor array was carefully designed based on tea polyphenol induced indicators displacement assay (IDA), using phenylboronic acids with different substituents as the receptors

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service