Skip to Content
Merck
All Photos(1)

Key Documents

434442

Supelco

Poly(styrene-co-divinylbenzene)

200-400 mesh particle size, 2 % cross-linked

Synonym(s):

Polystyrene crosslinked with divinylbenzene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[CH2CH(C6H5)]x[CH2CH[C6H4(CHCH2)]]y
CAS Number:
MDL number:
UNSPSC Code:
12000000
PubChem Substance ID:
NACRES:
SB.52

form

beads (fine)

crosslinking

2 % cross-linked

technique(s)

LPLC: suitable
thin layer chromatography (TLC): suitable

surface area

500 m2/g

matrix

Poly(styrene-co-divinylbenzene)

matrix active group

polymer

particle size

200-400 mesh

pore size

260 Å mean pore size

separation technique

reversed phase

SMILES string

C=Cc1ccccc1.C=Cc2ccc(C=C)cc2.C=Cc3cccc(C=C)c3.C=Cc4ccccc4C=C

InChI

1S/C10H12.C10H10.C8H8/c2*1-3-9-7-5-6-8-10(9)4-2;1-2-8-6-4-3-5-7-8/h3,5-8H,1,4H2,2H3;3-8H,1-2H2;2-7H,1H2

InChI key

NWUYHJFMYQTDRP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Poly(styrene-co-divinylbenzene) are crosslinked microspheres composed of various concentrations of divinylbenzene from 5 to 75mol% based on styrene monomer.[1] The gel-type PS beads are used as carrier to encapsulate metallocene catalysts.[2]

Application

Poly(styrene-co-divinylbenzene) may be used in separation media for TLC, paper and LPL chromatography and may also be used as adsorbent.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ethylene (Co) Polymerization with Metallocene Catalysts Encapsulated in Gel-Type Poly (styrene-co-divinylbenzene) Beads.
Hong, Sung Chul, Ursula Rief, and Marc Oliver Kristen.
Macromolecular Rapid Communications, 22, 1447-1454 (2001)
Fully crosslinked poly (styrene-co-divinylbenzene) microspheres by precipitation polymerization and their superior thermal properties.
Shim, Sang Eun, et al.
Journal of Polymer Science Part A: Polymer Chemistry, 42, 835-845 (2004)
T C Thomas et al.
American Industrial Hygiene Association journal, 41(8), 599-601 (1980-08-01)
An organic polymeric solid, Chromosorb 102, has been reported to collect various pesticides from air. In this study the collection efficiency of Chromosorb 102 for chlordane was evaluated. Preparation of sampling tubes containing this medium for field sampling is also
Sampling of airborne pesticides using Chromosorb 102.
T C Thomas et al.
Bulletin of environmental contamination and toxicology, 35(4), 460-465 (1985-10-01)
The use of Chromosorb 102 to air samples pre-concentration in the determination of low concentrated dibutylphthalate.
L Wolska
Bulletin of the Institute of Maritime and Tropical Medicine in Gdynia, 39(1-2), 81-89 (1988-01-01)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service