Skip to Content
Merck
All Photos(1)

Documents

433365

Sigma-Aldrich

Strontium sulfate

99%

Synonym(s):

Celestite

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
SrSO4
CAS Number:
Molecular Weight:
183.68
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:

Assay

99%

form

powder

mp

1606 °C (lit.)

solubility

aqueous acid: slightly soluble(lit.)
ethanol: insoluble(lit.)

density

3.96 g/mL at 25 °C (lit.)

SMILES string

[Sr++].[O-]S([O-])(=O)=O

InChI

1S/H2O4S.Sr/c1-5(2,3)4;/h(H2,1,2,3,4);/q;+2/p-2

InChI key

UBXAKNTVXQMEAG-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Alen V Gusa et al.
Environmental science & technology, 53(1), 373-382 (2018-12-12)
One of the major environmental concerns with the recovery of unconventional gas resource from Marcellus Shale is the presence of naturally occurring radioactive material (NORM) in produced water. Ra-226 is the major component of NORM with a half-life of 1600

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service