Skip to Content
Merck
All Photos(1)

Key Documents

426199

Sigma-Aldrich

Algal amino acid mixture-13C

98 atom % 13C

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
CZK 12,080.00

CZK 12,080.00

List priceCZK 60,400.00Save 80%

Please contact Customer Service for Availability

Request more information

Select a Size

Change View
1 G
CZK 12,080.00

About This Item

MDL number:
UNSPSC Code:
12352209
NACRES:
NA.12

CZK 12,080.00

List priceCZK 60,400.00Save 80%

Please contact Customer Service for Availability

Request more information

isotopic purity

98 atom % 13C

form

solid

technique(s)

bio NMR: suitable

Related Categories

Application

This product contains 16 amino acids extracted from a blue-green algal source. This product is deficient in glutamine, asparagine, cysteine, and tryptophan.

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Samuel Bryson et al.
The ISME journal, 11(12), 2781-2792 (2017-08-12)
Resource availability influences marine microbial community structure, suggesting that population-specific resource partitioning defines discrete niches. Identifying how resources are partitioned among populations, thereby characterizing functional guilds within the communities, remains a challenge for microbial ecologists. We used proteomic stable isotope

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service