Skip to Content
Merck

Skip To

424331

1-(3-Dimethylaminopropyl)-3-ethylcarbodiimide, polymer-bound

200-400 mesh, 2 % cross-linked with divinylbenzene

Synonym(s):

N-(3-Dimethylaminopropyl)-N′-ethylcarbodiimide polymer-bound, N-Ethyl-N′-(3-dimethylaminopropyl)carbodiimide polymer-bound, EDC polymer-bound

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
5 g
Please contact Customer Service for Availability
4 150,00 CZK

About This Item

MDL number:
UNSPSC Code:
12352111
PubChem Substance ID:
EC Number:
217-579-2
NACRES:
NA.22
Form:
powder

4 150,00 CZK


Please contact Customer Service for Availability

Request a Custom Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


form

powder

Quality Segment

crosslinking

2 % cross-linked with divinylbenzene

reaction suitability

reaction type: solution phase peptide synthesis, reactivity: carboxyl reactive

extent of labeling

0.5-1.5 mmol/g loading

particle size

200-400 mesh

SMILES string

CCN=C=NCCCN(C)C

InChI

1S/C8H17N3/c1-4-9-8-10-6-5-7-11(2)3/h4-7H2,1-3H3

InChI key

LMDZBCPBFSXMTL-UHFFFAOYSA-N

Application

Polymer-supported carbodiimide.


pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters