All Photos(1)
About This Item
Linear Formula:
F3C6H(NO2)NH2
CAS Number:
Molecular Weight:
192.10
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
Recommended Products
Assay
99%
mp
59-61 °C (lit.)
SMILES string
Nc1c(F)c(F)c(F)cc1[N+]([O-])=O
InChI
1S/C6H3F3N2O2/c7-2-1-3(11(12)13)6(10)5(9)4(2)8/h1H,10H2
InChI key
ILENVKAVEFKZSD-UHFFFAOYSA-N
General description
2,3,4-Trifluoro-6-nitroaniline is an aniline derivative. It is a building block used in chemical synthesis.
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service