Skip to Content
Merck
All Photos(1)

Key Documents

421960

Sigma-Aldrich

Bis(hexamethylene)triamine

high purity

Synonym(s):

6,6′-Iminodihexylamine, Bis(6-aminohexyl)amine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NH2(CH2)6NH(CH2)6NH2
CAS Number:
Molecular Weight:
215.38
Beilstein:
2323517
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor pressure

<0.01 mmHg ( 25 °C)

form

solid

refractive index

n20/D 1.49 (lit.)

bp

163-165 °C/4 mmHg (lit.)

mp

33-36 °C (lit.)

density

0.85 g/mL at 20 °C (lit.)
0.879 g/mL at 25 °C

SMILES string

NCCCCCCNCCCCCCN

InChI

1S/C12H29N3/c13-9-5-1-3-7-11-15-12-8-4-2-6-10-14/h15H,1-14H2

InChI key

MRNZSTMRDWRNNR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Bis(hexamethylene)triamine (Bis-HEA, BHMT, BHMTA) is an aliphatic polyamine. It is a spermidine analog and has been reported to lack the enhancing effect on mitochondrial Ca(2+) accumulation but shows inhibitory effect on the Ca(2+) and Pi-induced mitochondrial permeability transition.[1] Its ability in inhibiting shale swelling and dispersion in water-based drilling fluid has been investigated.[2]

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Dam. 1 - Skin Corr. 1A - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Co3O4-ZnO hierarchical nanostructures by electrospinning and hydrothermal methods.
Kanjwal MA, et al.
Applied Surface Science, 257(18), 7975-7981 (2011)
Antibacterial and photocatalytic properties of Ag/TiO2/ZnO nano-flowers prepared by facile one-pot hydrothermal process.
Pant HR, et al.
Ceramics International, 39(2), 1503-1510 (2013)
Bis (hexamethylene) triamine as potential shale inhibitor in water-based drilling fluid.
Zhong H, et al.
Open Petroleum Engineering Journal, 6, 49-56 (2013)
Interior synthesizing of ZnO nanoflakes inside nylon-6 electrospun nanofibers.
Panthi G, et al.
Journal of Applied Polymer Science, 127(3), 2025-2032 (2013)
Thomas McGlone et al.
Chemistry, an Asian journal, 4(10), 1612-1618 (2009-08-26)
The {W36} isopolyoxotungstate cluster provides a stable inorganic molecular platform for the binding of inorganic and organic guest molecules. This is achieved by a binding pocket formed by six terminal oxo ligands located in the central cavity of the all-inorganic

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service