Skip to Content
Merck
All Photos(1)

Documents

417491

Sigma-Aldrich

6-Methylchromone-2-carboxylic acid

98%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H8O4
CAS Number:
Molecular Weight:
204.18
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

98%

form

solid

mp

267-270 °C (lit.)

SMILES string

Cc1ccc2OC(=CC(=O)c2c1)C(O)=O

InChI

1S/C11H8O4/c1-6-2-3-9-7(4-6)8(12)5-10(15-9)11(13)14/h2-5H,1H3,(H,13,14)

InChI key

RKIDLZFIIVDZNX-UHFFFAOYSA-N

General description

6-Methylchromone-2-carboxylic acid is a chromone derivative. Chromones are cyclic compounds containing nine carbon atoms with sp2 hybridization. They are commonly referred as 1,4-benzapyranes or γ-benzopyrones. Its energy of combustion was measured by an isoperibolic micro-combustion calorimeter. Standard molar enthalpy of formation derived from the energy of combustion, for 6-methylchromone-2-carboxylic acid was reported to be -(656.2 ± 2.2)kJmol-1.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The standard molar enthalpies of formation of chromone-3-carboxylic acid, 6-methylchromone-2-carboxylic acid, and 6-methyl-4-chromanone determined by micro-combustion calorimetry.
Flores H, et al.
Journal of Thermal Analysis and Calorimetry, 117(1), 433-437 (2014)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service