Skip to Content
Merck
All Photos(1)

Documents

410330

Sigma-Aldrich

2,5-Dihydro-2,5-dimethoxyfurfuryl alcohol

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H12O4
CAS Number:
Molecular Weight:
160.17
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

97%

form

liquid

refractive index

n20/D 1.4590 (lit.)

bp

114 °C/14 mmHg (lit.)

density

1.164 g/mL at 25 °C (lit.)

SMILES string

COC1OC(CO)(OC)C=C1

InChI

1S/C7H12O4/c1-9-6-3-4-7(5-8,10-2)11-6/h3-4,6,8H,5H2,1-2H3

InChI key

ZYTSUANFIPZBDM-UHFFFAOYSA-N

General description

2,5-Dihydro-2,5-dimethoxyfurfuryl alcohol is a heterocyclic building block.

Application

2,5-Dihydro-2,5-dimethoxyfurfuryl alcohol may be used to investigate the bleomycin-induced alkali-labile DNA lesion.

Pictograms

Flame

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

50.0 °F - closed cup

Flash Point(C)

10 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chemistry of the bleomycin-induced alkali-labile DNA lesion.
Aso M, et al.
Journal of the American Chemical Society, 121(39), 9023-9033 (1999)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service