Skip to Content
Merck
All Photos(1)

Key Documents

3DXPLA014

Sigma-Aldrich

PLA 3D printing filament

white, diam. 2.85 mm

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
UNSPSC Code:
12352103

diam.

2.85 mm

color

white

Looking for similar products? Visit Product Comparison Guide

General description

3DXTech is made in the USA using 100% virgin resin and colorants - precision extruded onto 1kg reels (2.2 lbs.). The filament is then vacuum-sealed with desiccant to protect it from moisture.  

Preparation Note

Recommended Print Conditions: Extruder Temp: 190-220°C. Bed Temp: 23-60°C [No heat bed required]

Legal Information

Product of 3DXTech
3DXTech is a trademark of 3DXTech

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service