Skip to Content
Merck
All Photos(1)

Key Documents

398888

Sigma-Aldrich

Lead(II) zirconate

99% trace metals basis

Synonym(s):

Lead zirconate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
PbZrO3
CAS Number:
Molecular Weight:
346.42
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.23

Assay

99% trace metals basis

form

powder

impurities

<0.5% Hf

particle size

−325 mesh

SMILES string

[PbH2++].[O-][Zr]([O-])=O

InChI

1S/3O.Pb.Zr/q;2*-1;+2;

InChI key

RSCSNKLWUCUPDE-UHFFFAOYSA-N

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Repr. 1A - STOT RE 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

A significant limiting factor for wearable electronics and wireless sensors is the finite amount of energy that can be stored in on-board batteries.

Among various ceramics, one-dimensional (1-D) piezoelectric ceramics have attracted significant scientific attention for use in energy harvesting.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service