Skip to Content
Merck
All Photos(1)

Key Documents

38904

Sigma-Aldrich

1,3-Bis(cyanomethyl)imidazolium chloride

purum, ≥98.5% (HPLC/T)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H7ClN4
CAS Number:
Molecular Weight:
182.61
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

grade

purum

Assay

≥98.5% (HPLC/T)

form

powder

SMILES string

[Cl-].N#CCn1cc[n+](CC#N)c1

InChI

1S/C7H7N4.ClH/c8-1-3-10-5-6-11(7-10)4-2-9;/h5-7H,3-4H2;1H/q+1;/p-1

InChI key

PZSHHOPVYJHGQL-UHFFFAOYSA-M

General description

Nitrile-functionalized ionic liquid showing superior characteristics in Suzuki and Stille coupling reactions[1].

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Skin Irrit. 2 - STOT SE 3

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

N Parveen et al.
Soft matter, 13(10), 1988-1997 (2017-02-12)
Controlling the uptake of specific ions in polyelectrolyte multilayers is of interest for various fields of application. Here, we quantify the amount of cation of an ionic liquid, namely 1,3-bis(cyanomethyl)imidazolium chloride, incorporated into polyelectrolyte multilayers upon contact with an ionic
Dongbin Zhao et al.
Journal of the American Chemical Society, 126(48), 15876-15882 (2004-12-02)
A series of relatively low-cost ionic liquids, based on the N-butyronitrile pyridinium cation [C(3)CNpy](+), designed to improve catalyst retention, have been prepared and evaluated in Suzuki and Stille coupling reactions. Depending on the nature of the anion, these salts react

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service