Skip to Content
Merck
All Photos(1)

Documents

377708

Sigma-Aldrich

(Carbethoxyethylidene)triphenylphosphorane

94%

Synonym(s):

Ethyl 2-(triphenylphosphoranylidene)propionate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3C[=P(C6H5)3]CO2C2H5
CAS Number:
Molecular Weight:
362.40
EC Number:
MDL number:
UNSPSC Code:
12352108
PubChem Substance ID:
NACRES:
NA.22

grade

technical grade

Assay

94%

reaction suitability

reaction type: C-C Bond Formation

mp

158-162 °C (lit.)

storage temp.

2-8°C

SMILES string

CCOC(=O)C(C)=P(c1ccccc1)(c2ccccc2)c3ccccc3

InChI

1S/C23H23O2P/c1-3-25-23(24)19(2)26(20-13-7-4-8-14-20,21-15-9-5-10-16-21)22-17-11-6-12-18-22/h4-18H,3H2,1-2H3

InChI key

KZENFXVDPUMQOE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Other Notes

Contains triphenylphosphine oxide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service