Skip to Content
Merck
All Photos(1)

Key Documents

377228

Sigma-Aldrich

Ammonium lactate solution

technical grade, 20 wt. % in H2O

Synonym(s):

(±)-Ammonium lactate, Ammonium 2-hydroxypropionate

Sign Into View Organizational & Contract Pricing

Select a Size

100 ML
CZK 1,540.00

CZK 1,540.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
100 ML
CZK 1,540.00

About This Item

Linear Formula:
CH3CH(OH)CO2NH4
CAS Number:
Molecular Weight:
107.11
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.22

CZK 1,540.00


Please contact Customer Service for Availability

Request a Bulk Order

grade

technical grade

form

liquid

concentration

20 wt. % in H2O

impurities

2-3% lactic acid lactate, ammonium salt

refractive index

n20/D 1.363

density

1.054 g/mL at 25 °C

functional group

carboxylic acid
hydroxyl

SMILES string

N.CC(O)C(O)=O

InChI

1S/C3H6O3.H3N/c1-2(4)3(5)6;/h2,4H,1H3,(H,5,6);1H3

InChI key

RZOBLYBZQXQGFY-UHFFFAOYSA-N

General description

Ammonium lactate is an ammonium salt of lactic acid. It is generally used as a feedstock for ethyl lactate and as a component of moisturizers.[1][2][3]

Application

Ammonium lactate solution has been used in the ammonium lactate fermentation broth for the concentration of organic acid salts by conventional electrodialysis (EDC).[4]

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Marc Widenmeyer et al.
Nanomaterials (Basel, Switzerland), 10(4) (2020-04-23)
The determination of reversible phase transitions in the perovskite-type thermoelectric oxide Eu0.8Ca0.2TiO3-δ is fundamental, since structural changes largely affect the thermal and electrical transport properties. The phase transitions were characterized by heat capacity measurements, Rietveld refinements, and pair distribution function
Lorenza Rimassa et al.
Cancer treatment reviews, 77, 20-28 (2019-06-14)
Hepatocellular carcinoma (HCC) is the most common form of primary liver cancer. Sorafenib, regorafenib, lenvatinib and cabozantinib are tyrosine kinase inhibitors (TKIs) that target, in part, vascular endothelial growth factor receptors, and are approved in various regions of the world
Effect of solvent and impurity on synthesis of ethyl lactate from fermentation-derived ammonium lactate
Kasinathan P, et al.
Chemical Engineering Science, 66 (20), 4549-4554 (2011)
Effect of moisturizers on epidermal barrier function
Loden M
Clinics in Dermatology, 30 (3), 286-296 (2012)
Mesfin T Gebremikael et al.
Scientific reports, 6, 32862-32862 (2016-09-09)
The role of soil fauna in crucial ecosystem services such as nutrient cycling remains poorly quantified, mainly because of the overly reductionistic approach adopted in most experimental studies. Given that increasing nitrogen inputs in various ecosystems influence the structure and

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service