369594
2-(Trifluoromethyl)styrene
99%, contains 0.1% 4-tert-butylcatechol as inhibitor
Synonym(s):
1-(Trifluoromethyl)-2-vinylbenzene, 1-Ethenyl-2-(trifluoromethyl)benzene
About This Item
Recommended Products
Assay
99%
form
liquid
contains
0.1% 4-tert-butylcatechol as inhibitor
refractive index
n20/D 1.47 (lit.)
bp
61 °C/40 mmHg (lit.)
density
1.175 g/mL at 25 °C (lit.)
shipped in
wet ice
storage temp.
2-8°C
SMILES string
FC(F)(F)c1ccccc1C=C
InChI
1S/C9H7F3/c1-2-7-5-3-4-6-8(7)9(10,11)12/h2-6H,1H2
InChI key
VGWWQZSCLBZOGK-UHFFFAOYSA-N
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
3 - Flammable liquids
WGK
WGK 3
Flash Point(F)
107.6 °F - closed cup
Flash Point(C)
42 °C - closed cup
Personal Protective Equipment
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service