Recommended Products
Assay
97%
form
powder
mp
93-95 °C (lit.)
SMILES string
N#Cc1ccc2ccccc2n1
InChI
1S/C10H6N2/c11-7-9-6-5-8-3-1-2-4-10(8)12-9/h1-6H
InChI key
WDXARTMCIRVMAE-UHFFFAOYSA-N
General description
Mechanism of the photoinduced substitution reaction of 2-quinolinecarbonitrile in alcohols or ethers has been reported. Photoinitiated dimerization of 2-quinolinecarbonitrile in HCl-acidified 2-propanol/water has been investigated at 77K and 331K. Benzophenone-sensitization of 2-quinolinecarbonitrile has been reported to yield triazapentaphene.
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Photoinitiated radical-forming reactions of 2-quinolinecarbonitrile at 77 and 331 K.
The Journal of Organic Chemistry, 46(1), 34-38 (1981)
Biochimie, 125, 186-194 (2016-04-05)
Four cyclometalated iridium(III) complexes [Ir(dfppy)2(L)](+) (dfppy = 2-(2,4-difluorophenyl)pyridine, L = 6-(pyridin-2-yl)-1,3,5-triazine-2,4-diamine, Ir1; 6-(isoquinolin-1-yl)-1,3,5-triazine-2,4-diamine, Ir2; 6-(quinolin-2-yl)-1,3,5-triazine-2,4-diamine, Ir3; 6-(isoquinolin-3-yl)-1,3,5-triazine-2,4-diamine, Ir4) have been synthesized and characterized. Distinct from cisplatin, Ir1-Ir4 could specifically target mitochondria and induced apoptosis against various cancer cell lines, especially for cisplatin resistant cells.
The photochemistry of 2-quinolinecarbonitriles. III. The benzophenone-sensitized reaction.
Chemistry Letters (Jpn), 11, 1107-1110 (1975)
The Photochemistry of 2-Quinolinecarbonitriles. II. A Mechanism of the Photoinduced Substitution Reaction.
Bulletin of the Chemical Society of Japan, 47(4), 942-945 (1974)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service