Skip to Content
Merck
All Photos(2)

Key Documents

361534

Sigma-Aldrich

Nitromethanetrispropanol

Synonym(s):

4-(3-Hydroxypropyl)-4-nitro-1,7-heptanediol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
O2NC(CH2CH2CH2OH)3
CAS Number:
Molecular Weight:
235.28
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

mp

59-65 °C (lit.)

SMILES string

OCCCC(CCCO)(CCCO)[N+]([O-])=O

InChI

1S/C10H21NO5/c12-7-1-4-10(11(15)16,5-2-8-13)6-3-9-14/h12-14H,1-9H2

InChI key

WQOWCQQAYGOQNI-UHFFFAOYSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service