Skip to Content
Merck
All Photos(1)

Documents

348996

Sigma-Aldrich

2′-Benzyl-2,2-dimethylpropionanilide

99%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)3CCONHC6H4CH2C6H5
CAS Number:
Molecular Weight:
267.37
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

99%

mp

88-90 °C (lit.)

SMILES string

CC(C)(C)C(=O)Nc1ccccc1Cc2ccccc2

InChI

1S/C18H21NO/c1-18(2,3)17(20)19-16-12-8-7-11-15(16)13-14-9-5-4-6-10-14/h4-12H,13H2,1-3H3,(H,19,20)

InChI key

BDYMLDYIYIGVCY-UHFFFAOYSA-N

Application

2′-Benzyl-2,2-dimethylpropionanilide may be used in chemical synthesis.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service