Recommended Products
density
2.665 g/mL at 25 °C (lit.)
SMILES string
[K+].[K+].F[Si--](F)(F)(F)(F)F
InChI
1S/F6Si.2K/c1-7(2,3,4,5)6;;/q-2;2*+1
InChI key
UUHPPUWEDCVPCO-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
replaced by
Product No.
Description
Pricing
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral
Storage Class Code
6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
American journal of dentistry, 19(4), 248-252 (2006-08-31)
To evaluate the occluding ability of ammonium hexafluorosilicate (SiF). Dentin disks prepared from human extracted teeth were grouped as follows to prepare different situations of dentin hypersensitivity: (1) those sonicated for 20 minutes; (2) those treated with 0.5 mol/L EDTA
Dental materials : official publication of the Academy of Dental Materials, 24(2), 192-198 (2007-06-05)
Diamine silver fluoride [AgF: (NH(3))(2)AgF] has been used clinically in Japan, as it reduces dental caries and dentin hypersensitivity. However, AgF stains the teeth black due to silver precipitation. To overcome this drawback, the authors prepared ammonium hexafluorosilicate [SiF: (NH(4))(2)SiF(6)]
[Recurrent ventricular fibrillation in magnesium fluosilicate poisoning].
Medicina intensiva, 31(6), 344-346 (2007-08-01)
Forensic science international, 206(1-3), e20-e24 (2010-07-27)
Fluoride, of all inorganic substances, is among the least likely to be identified by a routine toxicological analysis. Acute poisonings with salts of hydrofluoric or fluorosilicic acid, however, although relatively uncommon, may occur. Some fluorosilicates, salts of fluorosilicic acid (e.g.
Caries research, 36(2), 136-141 (2002-05-31)
A previous study showed that a sodium hexafluorosilicate-calcium chloride-based two-solution fluoride (F) rinse containing 6 mmol/l of F was more effective than a 12 mmol/l F sodium fluoride rinse in depositing F on tooth surfaces and increasing oral F levels.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service