Skip to Content
Merck
All Photos(1)

Key Documents

334863

Sigma-Aldrich

2-Fluoro-4-nitroanisole

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
FC6H3(NO2)OCH3
CAS Number:
Molecular Weight:
171.13
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

mp

103-105 °C (lit.)

SMILES string

COc1ccc(cc1F)[N+]([O-])=O

InChI

1S/C7H6FNO3/c1-12-7-3-2-5(9(10)11)4-6(7)8/h2-4H,1H3

InChI key

XGMVTXUXZUPGGY-UHFFFAOYSA-N

General description

2-Fluoro-4-nitroanisole is a 2-halo-4-nitroanisole and its photoreaction with the nucleophiles hydroxide ion and pyridine has been investigated. Mechanism of photoreaction of 2-fluoro-4-nitroanisole with n-hexylamine has been investigated.

Application

2-Fluoro-4-nitroanisole was employed as alternative biochemical photoprobe for proteins.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gene G Wubbels et al.
The Journal of organic chemistry, 73(5), 1925-1934 (2008-01-25)
Photoreactions of 4-nitroanisole and the 2-halo-4-nitroanisoles (halogen = F, Cl, Br, and I) with the nucleophiles hydroxide ion and pyridine have been investigated quantitatively to extend the findings recently communicated for cyanide ion. The halonitroanisoles on excitation form triplet pi,pi*
The photosubstitution of 2-fluoro-4-nitroanisole with n-hexylamine. Evidence of two different triplet excited states in a dual mechanistic pathway.
Pleixats R and Marquet J.
Tetrahedron, 46(4), 1343-1352 (1990)
The Search for Biochemical Photoprobes. III. The Photoreactions of 4-Nitroveratrole and 2-Fluoro-4-nitroanisole with Bovine Pancreatic Ribonuclease A and with Model Nucleophiles.
Marquet J, et al.
Tetrahedron, 49(6), 1297-1306 (1993)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service