Skip to Content
Merck
All Photos(2)

Key Documents

334499

Sigma-Aldrich

Iron(III) fluoride trihydrate

Synonym(s):

Ferric fluoride trihydrate, Iron trifluoride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
FeF3 · 3H2O
CAS Number:
Molecular Weight:
166.89
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23

form

powder

concentration

32.3-34.6% Fe (by Na2S2O3, titration)

application(s)

battery manufacturing

SMILES string

O.O.O.F[Fe](F)F

InChI

1S/3FH.Fe.3H2O/h3*1H;;3*1H2/q;;;+3;;;/p-3

InChI key

XILAUHRDSUSGOG-UHFFFAOYSA-K

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Lithium-ion batteries' characteristics make them popular for electricity storage due to portability, rechargeability, and low cost.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service