Skip to Content
Merck
All Photos(1)

Documents

333395

Sigma-Aldrich

Dichlorosilane

≥99.99% trace metals basis

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Cl2SiH2
CAS Number:
Molecular Weight:
101.01
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:

vapor density

3.5 (vs air)

vapor pressure

1254 mmHg ( 20 °C)

description

film resistivity > 50 Ω-cm

Assay

≥99.99% trace metals basis

autoignition temp.

136 °F

expl. lim.

99 %

bp

8.3 °C (lit.)

mp

−122 °C (lit.)

SMILES string

Cl[SiH2]Cl

InChI

1S/Cl2H2Si/c1-3-2/h3H2

InChI key

MROCJMGDEKINLD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Dichlorosilane (DCS) is a halogenated silicon based precursor that facilitates the growth of a variety of silane based films such as silicon oxide, silicon nitride, silicon carbide, and epitaxial growth of silica.

Application

DCS is widely used in the synthesis of a variety of silicon-based materials, which find applications in the development of organic electronics based devices, including OFETs, MEMS, NEMS, MOSFETS, and lithium-ion batteries.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Inhalation - Eye Dam. 1 - Flam. Gas 1 - Press. Gas Liquefied gas - Skin Corr. 1B

Supplementary Hazards

Storage Class Code

2A - Gases

WGK

WGK 1

Flash Point(F)

-34.6 °F

Flash Point(C)

-37 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J Komatsu et al.
Journal of biomolecular structure & dynamics, 22(3), 331-337 (2004-10-12)
This report shows a new DNA stretching method using migration of an ice-water interface. DNA molecules were stretched accompanying the migration of the solid-liquid interface and immobilized in frozen area. This simple method needs no chemical modification to keep DNA
T Chen et al.
Electrophoresis, 20(12), 2412-2419 (1999-09-28)
The feasibility of polymeric phases based on a silicone polymer backbone as pseudostationary phases for electrokinetic chromatography has been investigated. Silicone phases were studied because of the range of chemistries that could be developed based on these backbones, and because
Low pressure chemical vapor deposition of silicon carbide from dichlorosilane and acetylene.
Wang C and Tsai D
Materials Chemistry and Physics, 63(3), 196-201 (2000)
Dichlorosilane-derived nano-silicon inside hollow carbon spheres as a high-performance anode for Li-ion batteries.
Jaumann T, et al.
Journal of Material Chemistry C, 5(19), 9262-9271 (2017)
Stress control of polycrystalline 3C−SiC films in a large-scale LPCVD reactor using 1, 3-disilabutane and dichlorosilane as precursors.
Roper CS, et al.
Journal of Micromechanics and Microengineering, 16(12), 2736-2736 (2006)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service