Skip to Content
Merck
All Photos(1)

Key Documents

331856

Sigma-Aldrich

(2S,3R)-(+)-N-Z-6-oxo-2,3-diphenylmorpholine

99%

Synonym(s):

Benzyl (2S,3R)-(+)-6-oxo-2,3-diphenyl-4-morpholinecarboxylate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H21NO4
CAS Number:
Molecular Weight:
387.43
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

optical activity

[α]25/D +66°, c = 5.5 in methylene chloride

mp

205-207 °C (lit.)

storage temp.

2-8°C

SMILES string

O=C1CN([C@@H]([C@@H](O1)c2ccccc2)c3ccccc3)C(=O)OCc4ccccc4

InChI

1S/C24H21NO4/c26-21-16-25(24(27)28-17-18-10-4-1-5-11-18)22(19-12-6-2-7-13-19)23(29-21)20-14-8-3-9-15-20/h1-15,22-23H,16-17H2/t22-,23+/m1/s1

InChI key

HECRUWTZAMPQOS-PKTZIBPZSA-N

Application

(2S,3R)-(+)-N-Z-6-oxo-2,3-diphenylmorpholine can be used as a starting material for the synthesis of:
  • α-Amino-β-silyloxy-ester, a key intermediate for the preparation of antimalarial drug quinine and its analogs.
  • R, R-Formylglycine dimethylacetal, a key intermediate for the preparation of tuberculostatic compound capreomycin IB.
  • 2R,5R,6S-2-(Methoxycarbonylmethyl)-5,6-diphenylmorpholine hydrochloride, a key intermediate for the preparation of an antibiotic (+)-negamycin.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Asymmetric synthesis of (+)-negamycin
Jain RP and Williams RM
The Journal of Organic Chemistry, 67(18), 6361-6365 (2002)
Asymmetric synthesis of (2S, 3R)-capreomycidine and the total synthesis of capreomycin IB
DeMong DE and Williams RM
Journal of the American Chemical Society, 125(28), 8561-8565 (2003)
Synthetic studies on quinine: quinuclidine construction via a Ketone Enolate regio-and diastereoselective Pd-mediated allylic alkylation
Johns DM, et al.
Organic Letters, 8(18), 4051-4054 (2006)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service