Skip to Content
Merck
All Photos(1)

Documents

331821

Sigma-Aldrich

tert-Butyl 6-oxo-2,3-diphenyl-4-morpholinecarboxylate

≥99%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H23NO4
CAS Number:
Molecular Weight:
353.41
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

≥99%

mp

191-193 °C (lit.)

storage temp.

2-8°C

SMILES string

CC(C)(C)OC(=O)N1CC(=O)OC(C1c2ccccc2)c3ccccc3

InChI

1S/C21H23NO4/c1-21(2,3)26-20(24)22-14-17(23)25-19(16-12-8-5-9-13-16)18(22)15-10-6-4-7-11-15/h4-13,18-19H,14H2,1-3H3

InChI key

MRUKRSQUUNYOFK-UHFFFAOYSA-N

Application

tert-Butyl-(2R,3S)-(−)-6-oxo-2,3-diphenyl-4-morpholinecarboxylate was used in asymmetric synthesis of unnatural amino acids.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

R M Williams
Methods in molecular medicine, 23, 339-356 (1999-01-01)
Unnatural and naturally occurring nonproteinogenic α-amino acids have become important building blocks for the synthesis of biologically active peptides and peptidomimetic drug molecules. The asymmetric synthesis of α-amino acids has therefore become quite important as an indispensable research tool in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service