Skip to Content
Merck
All Photos(1)

Key Documents

323624

Sigma-Aldrich

2′,3′-Dideoxyuridine

98%

Synonym(s):

ddU

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H12N2O4
CAS Number:
Molecular Weight:
212.20
Beilstein:
750592
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

optical activity

[α]20/D +34°, c = 0.4 in H2O

mp

127-129 °C (lit.)

functional group

ether
hydroxyl

SMILES string

OC[C@@H]1CC[C@@H](O1)N2C=CC(=O)NC2=O

InChI

1S/C9H12N2O4/c12-5-6-1-2-8(15-6)11-4-3-7(13)10-9(11)14/h3-4,6,8,12H,1-2,5H2,(H,10,13,14)/t6-,8+/m0/s1

InChI key

BTOTXLJHDSNXMW-POYBYMJQSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Research tool for antiviral and anticancer studies.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

H Shirae et al.
Applied and environmental microbiology, 55(2), 419-424 (1989-02-01)
A novel microbial method for the production of 2',3'-dideoxynucleosides by transdideoxyribosylation has been developed. By screening microorganisms producing 2',3'-dideoxyadenosine (DDA) from 2',3'-dideoxyuridine (DDU) and adenine, Escherichia coli AJ 2595 was selected as the best producer. Optimal pH and temperature for
Z Hao et al.
Molecular pharmacology, 37(2), 157-163 (1990-02-01)
2',3'-Dideoxyuridine (ddUrd) exhibits poor if any anti-human immunodeficiency virus (HIV) activity in ATH8 and MT-4 cells. This is in agreement with the failure of ddUrd to be efficiently anabolized intracellularly to its 5'-triphosphate metabolite. However, 2',3'-dideoxyuridine-5'-triphosphate (ddUTP) proved to be
Evelina Colacino et al.
Nucleosides, nucleotides & nucleic acids, 22(11), 2013-2026 (2003-12-19)
In this article, we describe the synthesis of 5-nitro-1-(2-deoxy-alpha-D-erythro-pentofuranosyl)cytosine (4alpha), 5-nitro-1-(2-deoxy-beta-D-erythro-pentofuranosyl)cytosine (4beta), 5-amino-1-(2-deoxy-alpha-D-erythro-pentofuranosyl)cytosine (5alpha), 5-nitro-1-(2-deoxy-beta-D-erythro-pentofuranosyl)cytosine (5beta), 5-nitro-1-(2,3-dideoxy-beta-D-ribofuranosyl)uracil (6beta), 5-amino-1-(2,3-dideoxy-alpha,beta-D-ribofuranosyl)uracil (7), 5-nitro-1-(2,3-dideoxy-alpha,beta-D-ribofuranosyl)cytosine (8) and 5-amino-1-(2,3-dideoxy-beta-D-ribofuranosyl)cytosine (9beta). The prepared compounds were tested for their activity against HIV and HBV viruses, but they
Youcef Mehellou et al.
Bioorganic & medicinal chemistry letters, 17(13), 3666-3669 (2007-05-09)
We report the synthesis of 2',3'-didehydro-2',3'-dideoxyuridine (d4U) and 2',3'-dideoxyuridine (ddU) phosphoramidate 'ProTide' derivatives and their evaluation against HIV-1 and HIV-2. In addition, we conducted molecular modeling studies on both d4U and ddU monophosphates to investigate their second phosphorylation process. The
C McGuigan et al.
FEBS letters, 351(1), 11-14 (1994-08-29)
As part of our effort to deliver masked phosphates inside living cells we have discovered that certain phosphate triester derivatives of the inactive nucleoside analogue, dideoxy uridine (ddU) are inhibitors of HIV replication at microM levels. Moreover, we note that

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service