Skip to Content
Merck
All Photos(1)

Documents

319724

Sigma-Aldrich

Perfluoro-1-octanesulfonyl fluoride

95% (sum of isomers)

Synonym(s):

Heptadecafluorooctanesulfonyl fluoride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CF3(CF2)7SO2F
CAS Number:
Molecular Weight:
502.12
Beilstein:
1718067
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor density

>1 (vs air)

vapor pressure

<10 mmHg ( 20 °C)

Assay

95% (sum of isomers)

form

liquid

reaction suitability

reaction type: click chemistry

refractive index

n20/D 1.301 (lit.)

bp

154-155 °C (lit.)

density

1.824 g/mL at 25 °C (lit.)

SMILES string

FC(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)S(F)(=O)=O

InChI

1S/C8F18O2S/c9-1(10,3(13,14)5(17,18)7(21,22)23)2(11,12)4(15,16)6(19,20)8(24,25)29(26,27)28

InChI key

BHFJBHMTEDLICO-UHFFFAOYSA-N

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 2 - Carc. 2 - Lact. - Repr. 1B - STOT RE 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service