Skip to Content
Merck
All Photos(2)

Documents

310913

Sigma-Aldrich

7-Bromo-1-heptanol

95%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Br(CH2)7OH
CAS Number:
Molecular Weight:
195.10
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

95%

form

liquid

refractive index

n20/D 1.482 (lit.)

bp

111-112 °C/4 mmHg (lit.)

density

1.269 g/mL at 25 °C (lit.)

SMILES string

OCCCCCCCBr

InChI

1S/C7H15BrO/c8-6-4-2-1-3-5-7-9/h9H,1-7H2

InChI key

MMXRRNUXCHUHOE-UHFFFAOYSA-N

Application

7-Bromo-1-heptanol was used in the synthesis of (9Z,12E)-[1-13C]-octadeca-9,12-dienoic acid and(9Z,12Z,15E)-[1-13C]-octadeca-9,12,15-trienoic acid.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

O Loreau et al.
Chemistry and physics of lipids, 106(1), 65-78 (2000-07-06)
Several grams of labelled trans linoleic and linolenic acids with high chemical and isomeric purities (>97%) have been prepared for human metabolism studies. A total of 12.5 g of (9Z, 12E)-[1-(13)C]-octadeca-9,12-dienoic acid and 6.3 g of (9Z,12Z, 15E)-[1-(13)C]-octadeca-9,12,15-trienoic acid were

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service