Skip to Content
Merck
All Photos(1)

Key Documents

310425

Sigma-Aldrich

(−)-Camphene

technical grade, 75% (90% as camphene and fenchene)

Synonym(s):

(1S)-2,2-Dimethyl-3-methylenebicyclo[2.2.1]heptane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H16
CAS Number:
Molecular Weight:
136.23
Beilstein:
2204256
EC Number:
MDL number:
UNSPSC Code:
12352002
PubChem Substance ID:
NACRES:
NA.22

grade

technical grade

Assay

75% (90% as camphene and fenchene)

optical activity

[α]20/D −48°, c = 4 in ethanol
[α]25/D -35 to -43°, c = 4 in ethanol

bp

157.5-158.5 °C (lit.)

mp

36-38 °C (lit.)

density

0.84 g/mL at 25 °C (lit.)

SMILES string

CC1(C)[C@H]2CC[C@H](C2)C1=C

InChI

1S/C10H16/c1-7-8-4-5-9(6-8)10(7,2)3/h8-9H,1,4-6H2,2-3H3/t8-,9+/m1/s1

InChI key

CRPUJAZIXJMDBK-BDAKNGLRSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Flame

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Sol. 1

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

WGK 3

Flash Point(F)

93.2 °F - closed cup

Flash Point(C)

34 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service