Skip to Content
Merck
All Photos(1)

Documents

30905

Sigma-Aldrich

Decontamination solution

Synonym(s):

Polyethylene glycol ethanol

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352103

technique(s)

protein quantification: suitable

density

1.02 g/mL at 20 °C

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

73.4 °F - closed cup

Flash Point(C)

23 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service