Skip to Content
Merck
All Photos(1)

Key Documents

308781

Sigma-Aldrich

N,N-Dimethylformamide-d7

99.5 atom % D

Synonym(s):

DMF-d7, Heptadeutero-N,N-dimethylformamide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
DCON(CD3)2
CAS Number:
Molecular Weight:
80.14
Beilstein:
1908468
MDL number:
UNSPSC Code:
12142201
PubChem Substance ID:

isotopic purity

99.5 atom % D

packaging

pk of 5 × 0.5 mL

refractive index

n20/D 1.428 (lit.)

bp

153 °C (lit.)

density

1.03 g/mL at 25 °C (lit.)

mass shift

M+7

SMILES string

[2H]C(=O)N(C([2H])([2H])[2H])C([2H])([2H])[2H]

InChI

1S/C3H7NO/c1-4(2)3-5/h3H,1-2H3/i1D3,2D3,3D

InChI key

ZMXDDKWLCZADIW-YYWVXINBSA-N

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Eye Irrit. 2 - Flam. Liq. 3 - Repr. 1B

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

135.5 °F - closed cup

Flash Point(C)

57.5 °C - closed cup


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH SVHC Candidate List

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service