Skip to Content
Merck
All Photos(2)

Key Documents

307785

Sigma-Aldrich

Ethyl undecylenate

97%

Synonym(s):

Ethyl 10-undecenoate

Sign Into View Organizational & Contract Pricing

Select a Size

100 ML
CZK 1,500.00

CZK 1,500.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
100 ML
CZK 1,500.00

About This Item

Linear Formula:
H2C=CH(CH2)8CO2C2H5
CAS Number:
Molecular Weight:
212.33
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

CZK 1,500.00


Please contact Customer Service for Availability

Request a Bulk Order

Assay

97%

form

liquid

refractive index

n20/D 1.439 (lit.)

bp

258-259 °C/761 mmHg (lit.)

density

0.879 g/mL at 25 °C (lit.)

functional group

allyl
ester

SMILES string

CCOC(=O)CCCCCCCCC=C

InChI

1S/C13H24O2/c1-3-5-6-7-8-9-10-11-12-13(14)15-4-2/h3H,1,4-12H2,2H3

InChI key

FXNFFCMITPHEIT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Ethyl undecylenate was used to prepare mixed monolayers by undergoing photochemical reaction with hydrogen-terminated silicon[1].

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yong-Jun Liu et al.
Langmuir : the ACS journal of surfaces and colloids, 20(10), 4039-4050 (2005-06-23)
The structure, reactivity, and acid-base properties of mixed monolayers prepared by photochemical reaction of hydrogen-terminated silicon with mixtures of ethyl undecylenate and n-alkenes were studied by ATR-FTIR spectroscopy and contact-angle measurements. The surface composition of the mixed monolayers and its

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service