Skip to Content
Merck
All Photos(1)

Key Documents

300519

Sigma-Aldrich

(2R)-(−)-Glycidyl tosylate

99%

Synonym(s):

(R)-(−)-Glycidyl p-toluenesulfonate, (R)-(−)-Glycidyl tosylate, (R)-(−)-Oxirane-2-methanol p-toluenesulfonate salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H12O4S
CAS Number:
Molecular Weight:
228.26
Beilstein:
3592142
MDL number:
UNSPSC Code:
12352000

Assay

99%

optical activity

[α]19/D −17°, c = 2.75 in chloroform

mp

46-49 °C (lit.)

SMILES string

Cc1ccc(cc1)S(=O)(=O)OC[C@H]2CO2

InChI

1S/C10H12O4S/c1-8-2-4-10(5-3-8)15(11,12)14-7-9-6-13-9/h2-5,9H,6-7H2,1H3/t9-/m1/s1

InChI key

NOQXXYIGRPAZJC-SECBINFHSA-N

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Carc. 1B - Eye Dam. 1 - Muta. 2 - Skin Sens. 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

>230.0 °F

Flash Point(C)

> 110 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service