Skip to Content
Merck
All Photos(1)

Documents

299588

Sigma-Aldrich

(3-Trimethylsilyl-2-propynyl)triphenylphosphonium bromide

98%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H26BrPSi
CAS Number:
Molecular Weight:
453.43
Beilstein:
3581358
MDL number:
UNSPSC Code:
12352003
PubChem Substance ID:

Assay

98%

reaction suitability

reaction type: C-C Bond Formation

mp

166-167 °C (dec.) (lit.)

SMILES string

[Br-].C[Si](C)(C)C#CC[P+](c1ccccc1)(c2ccccc2)c3ccccc3

InChI

1S/C24H26PSi.BrH/c1-26(2,3)21-13-20-25(22-14-7-4-8-15-22,23-16-9-5-10-17-23)24-18-11-6-12-19-24;/h4-12,14-19H,20H2,1-3H3;1H/q+1;/p-1

InChI key

PBSHVEOONSKWJF-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application

Reactant for:
  • Horner-Wadsworth-Emmons olefination
  • Aromatic ring umpolung
  • Stereoselective Wittig reaction
  • Bromoetherification reactions
  • Olefination of Garner′s aldehyde
  • Regioselective Wittig reaction

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service